missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzophenone, 97%

Catalog No. p-156951
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125740250 25 g Glass bottle
AC125741000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC125740250 Supplier Thermo Scientific Chemicals Supplier No. 125740250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-94-9 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-94-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00008403
InChI Key SWFHGTMLYIBPPA-UHFFFAOYSA-N
Synonym 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone
PubChem CID 69146
IUPAC Name (4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 58°C to 63°C
Color Orange-Yellow to White
Boiling Point 354°C to 356°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COC6H5
Quantity 25 g
Beilstein 08, 159
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol and ether
Formula Weight 212.25
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxybenzophenone, 97%
Show More Show Less
EINECSNumber 210-285-5
RTECSNumber PC4962500

RUO – Research Use Only