Learn More
4-Methoxybenzonitrile, 99%
CAS: 874-90-8 | C8H7NO | 133.15 g/mol
$76.26 - $529.59
Chemical Identifiers
| CAS | 874-90-8 |
|---|---|
| Molecular Formula | C8H7NO |
| Molecular Weight (g/mol) | 133.15 |
| MDL Number | MFCD00001818 |
| InChI Key | XDJAAZYHCCRJOK-UHFFFAOYSA-N |
| Synonym | anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile |
| PubChem CID | 70129 |
| IUPAC Name | 4-methoxybenzonitrile |
| SMILES | COC1=CC=C(C=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1594506
|
Thermo Scientific Chemicals
A1594506 |
5 g |
Each for $76.26
|
|
|||||
|
AAA1594514
|
Thermo Scientific Chemicals
A1594514 |
25 g |
Each for $195.95
|
|
|||||
|
AAA1594522
|
Thermo Scientific Chemicals
A1594522 |
100 g |
Each for $529.59
|
|
|||||
Description
4-Methoxybenzonitrile is used as raw material of parfum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzonitrile is used as raw material of parfum.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Chemical Identifiers
| 874-90-8 | |
| 133.15 | |
| XDJAAZYHCCRJOK-UHFFFAOYSA-N | |
| 70129 | |
| COC1=CC=C(C=C1)C#N |
| C8H7NO | |
| MFCD00001818 | |
| anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile | |
| 4-methoxybenzonitrile |
Specifications
| 874-90-8 | |
| 256°C to 257°C | |
| C8H7NO | |
| 5 g | |
| 508117 | |
| Insoluble in water. | |
| COC1=CC=C(C=C1)C#N | |
| 133.15 | |
| 133.15 | |
| 4-Methoxybenzonitrile |
| 57°C to 61°C | |
| Almond-like | |
| MFCD00001818 | |
| UN3439 | |
| anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile | |
| XDJAAZYHCCRJOK-UHFFFAOYSA-N | |
| 4-methoxybenzonitrile | |
| 70129 | |
| 99% |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 212-871-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only