missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoic Anhydride 97.0+%, TCI America™

Catalog No. p-7132944
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19735G 5 g
M197325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19735G Supplier TCI America Supplier No. M19735G
Only null left

Chemical Identifiers

CAS 794-94-5
Molecular Formula C16H14O5
Molecular Weight (g/mol) 286.283
MDL Number MFCD00017175
InChI Key YGMHIBLUWGDWKP-UHFFFAOYSA-N
Synonym 4-methoxybenzoic anhydride, p-anisic anhydride, p-anisic acid anhydride, benzoic acid, 4-methoxy-, anhydride, p-methoxybenzoic anhydride, 4-anisic anhydride, 4-methoxybenzoic acid anhydride, 4-methoxybenzoyl 4-methoxybenzoate, 4-methoxyphenyl anhydride, anisic anhydride
PubChem CID 69928
IUPAC Name (4-methoxybenzoyl) 4-methoxybenzoate
SMILES COC1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)OC

Specifications

Melting Point 98°C
Color White-Yellow
Boiling Point 310°C
Quantity 5 g
Formula Weight 286.28
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzoic Anhydride
RTECSNumber BZ5300000
TSCA No