missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoic anhydride, 98%

Catalog No. AAL0618006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0618006 5 g
AAL0618014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0618006 Supplier Thermo Scientific Chemicals Supplier No. L0618006

CAS: 794-94-5 | C16H14O5 | 286.283 g/mol

4-Methoxybenzoic anhydride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzoic anhydride is used as pharmaceutical intermediates.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Moisture Sensitive. Protect from humidity and water. Incompatible with moisture and oxidizing agents.

Chemical Identifiers

CAS 794-94-5
Molecular Formula C16H14O5
Molecular Weight (g/mol) 286.283
MDL Number MFCD00017175
InChI Key YGMHIBLUWGDWKP-UHFFFAOYSA-N
Synonym 4-methoxybenzoic anhydride, p-anisic anhydride, p-anisic acid anhydride, benzoic acid, 4-methoxy-, anhydride, p-methoxybenzoic anhydride, 4-anisic anhydride, 4-methoxybenzoic acid anhydride, 4-methoxybenzoyl 4-methoxybenzoate, 4-methoxyphenyl anhydride, anisic anhydride
PubChem CID 69928
IUPAC Name (4-methoxybenzoyl) 4-methoxybenzoate
SMILES COC1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)OC

Specifications

Melting Point 95°C to 99°C
Quantity 5 g
Beilstein 545712
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohol,ether. Insoluble in water.
Formula Weight 286.29
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzoic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-345-6
RTECSNumber BZ5300000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only