missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoic acid, 98+%

CAS: 100-09-4 | C8H8O3 | 152.15 g/mol

Supplier:  Thermo Scientific Chemicals A156970B

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA156970B

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

4-Methoxybenzoic acid is used in oxidation and reduction of cytochrome c in solution through different self-assembled monolayers on gold electrodes using cyclic voltammetry. p-Anisic acid has antiseptic properties. It is also used as an intermediate in the preparation of more complex organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzoic acid is used in oxidation and reduction of cytochrome c in solution through different self-assembled monolayers on gold electrodes using cyclic voltammetry. p-Anisic acid has antiseptic properties. It is also used as an intermediate in the preparation of more complex organic compounds.

Solubility
Soluble in water at 20°C 0.3g/L. Soluble in alcohol, ethyl acetate and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Spécifications

Spécifications

4-Methoxybenzoic acid
182°C to 185°C
3 to 4
185°C (365°F)
C8H8O3
1000 g
14,666
Soluble in water at 20°C 0.3g/L. Soluble in alcohol,ethyl acetate and ether.
COC1=CC=C(C=C1)C(O)=O
152.15
CHEBI:40813
≥98%
100-09-4
1.385
275°C to 277°C
Odorless
MFCD00002542
508910
p-anisic acid, anisic acid, p-methoxybenzoic acid, draconic acid, 4-anisic acid, benzoic acid, 4-methoxy, anisic acid, para, 4-methoxybenzoate, para-anisic acid, anisic acid, p-isomer
ZEYHEAKUIGZSGI-UHFFFAOYSA-N
4-methoxybenzoic acid
7478
152.15

Identifiants chimiques

100-09-4
152.15
ZEYHEAKUIGZSGI-UHFFFAOYSA-N
7478
4-methoxybenzoic acid
C8H8O3
MFCD00002542
p-anisic acid, anisic acid, p-methoxybenzoic acid, draconic acid, 4-anisic acid, benzoic acid, 4-methoxy, anisic acid, para, 4-methoxybenzoate, para-anisic acid, anisic acid, p-isomer
CHEBI:40813
COC1=CC=C(C=C1)C(O)=O
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 202-818-5

missing translation for 'rtecsNumber' : BZ4395000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only