missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoic acid, 98+%

Catalog No. AAA156970I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA156970I 5000 g
AAA1569730 250 g
AAA156970B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA156970I Supplier Thermo Scientific Chemicals Supplier No. A156970I
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-09-4 | C8H8O3 | 152.15 g/mol

4-Methoxybenzoic acid is used in oxidation and reduction of cytochrome c in solution through different self-assembled monolayers on gold electrodes using cyclic voltammetry. p-Anisic acid has antiseptic properties. It is also used as an intermediate in the preparation of more complex organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzoic acid is used in oxidation and reduction of cytochrome c in solution through different self-assembled monolayers on gold electrodes using cyclic voltammetry. p-Anisic acid has antiseptic properties. It is also used as an intermediate in the preparation of more complex organic compounds.

Solubility
Soluble in water at 20°C 0.3g/L. Soluble in alcohol, ethyl acetate and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 100-09-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002542
InChI Key ZEYHEAKUIGZSGI-UHFFFAOYSA-N
Synonym p-anisic acid, anisic acid, p-methoxybenzoic acid, draconic acid, 4-anisic acid, benzoic acid, 4-methoxy, anisic acid, para, 4-methoxybenzoate, para-anisic acid, anisic acid, p-isomer
PubChem CID 7478
ChEBI CHEBI:40813
IUPAC Name 4-methoxybenzoic acid
SMILES COC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 182°C to 185°C
Density 1.385
pH 3 to 4
Boiling Point 275°C to 277°C
Flash Point 185°C (365°F)
Odor Odorless
Quantity 5000 g
Beilstein 508910
Merck Index 14,666
Solubility Information Soluble in water at 20°C 0.3g/L. Soluble in alcohol,ethyl acetate and ether.
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzoic acid
Show More Show Less
EINECSNumber 202-818-5
RTECSNumber BZ4395000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only