missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenethiol 96.0+%, TCI America™

Catalog No. p-7132667
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13335G 5 g
M133325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13335G Supplier TCI America Supplier No. M13335G
Only null left

Chemical Identifiers

CAS 696-63-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004849
InChI Key NIFAOMSJMGEFTQ-UHFFFAOYSA-N
Synonym 4-methoxythiophenol, p-methoxybenzenethiol, benzenethiol, 4-methoxy, 4-methoxybenzene-1-thiol, benzenethiol, p-methoxy, 4-mercaptoanisole, 4-methoxy thiophenol, 4-methoxy-benzenethiol, para-methoxybenzenethiol, 4-mercaptoanisol
PubChem CID 12765
IUPAC Name 4-methoxybenzenethiol
SMILES COC1=CC=C(C=C1)S

Specifications

Color Yellow
Boiling Point 227°C
Quantity 5 g
UN Number 2810
Formula Weight 140.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzenethiol
RTECSNumber DC1800000
TSCA No