missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America M0802100G
Specifications
| 4-Methoxybenzenesulfonyl Chloride | |
| White-Yellow | |
| C7H7ClO3S | |
| 100 g | |
| 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy | |
| COC1=CC=C(C=C1)S(Cl)(=O)=O | |
| 206.64 | |
| 206.64 | |
| Crystalline Lumps |
| 98-68-0 | |
| 173°C | |
| MFCD00007446 | |
| 3261 | |
| DTJVECUKADWGMO-UHFFFAOYSA-N | |
| 4-methoxybenzene-1-sulfonyl chloride | |
| 7401 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 98-68-0 | |
| 206.64 | |
| DTJVECUKADWGMO-UHFFFAOYSA-N | |
| 7401 | |
| COC1=CC=C(C=C1)S(Cl)(=O)=O |
| C7H7ClO3S | |
| MFCD00007446 | |
| 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy | |
| 4-methoxybenzene-1-sulfonyl chloride |
Safety and Handling
EINECSNumber : (9)-0053
TSCA : No