missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7132369
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M080225G 25 g
M0802100G 100 g
M0802500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M080225G Supplier TCI America Supplier No. M080225G
Only null left

Chemical Identifiers

CAS 98-68-0
Molecular Formula C7H7ClO3S
Molecular Weight (g/mol) 206.64
MDL Number MFCD00007446
InChI Key DTJVECUKADWGMO-UHFFFAOYSA-N
Synonym 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy
PubChem CID 7401
IUPAC Name 4-methoxybenzene-1-sulfonyl chloride
SMILES COC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Color White-Yellow
Boiling Point 173°C
Quantity 25 g
UN Number 3261
Formula Weight 206.64
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methoxybenzenesulfonyl Chloride
EINECSNumber (9)-0053
TSCA No