Learn More
4-Methoxybenzenesulfonyl chloride, 98+%
CAS: 98-68-0 | C7H7ClO3S | 206.64 g/mol
Supplier: Thermo Scientific Chemicals A1378814
Description
4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.
Solubility
Soluble in Toluene. Decomposes in water.
Notes
Moisture Sensitive. Store at 4°C. Store in a cool and dark place. Protect from moisture. Store away from strong bases.
Specifications
| 4-Methoxybenzenesulfonyl chloride | |
| 37°C to 42°C | |
| >110°C (230°F) | |
| MFCD00007446 | |
| UN3261 | |
| Moisture sensitive | |
| Soluble in Toluene. Decomposes in water. | |
| COC1=CC=C(C=C1)S(Cl)(=O)=O | |
| 206.64 | |
| 206.65 |
| 98-68-0 | |
| 119°C to 122°C (0.4 mmHg) | |
| C7H7ClO3S | |
| 25 g | |
| 609005 | |
| 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy | |
| DTJVECUKADWGMO-UHFFFAOYSA-N | |
| 4-methoxybenzenesulfonyl chloride | |
| 7401 | |
| ≥98% |
Chemical Identifiers
| 98-68-0 | |
| 206.64 | |
| DTJVECUKADWGMO-UHFFFAOYSA-N | |
| 7401 | |
| COC1=CC=C(C=C1)S(Cl)(=O)=O |
| C7H7ClO3S | |
| MFCD00007446 | |
| 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy | |
| 4-methoxybenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 202-692-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only