missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenesulfonyl chloride, 98+%

Catalog No. p-7021453
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1378814 25 g
AAA1378822 100 g
AAA1378836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1378814 Supplier Thermo Scientific Chemicals Supplier No. A1378814

CAS: 98-68-0 | C7H7ClO3S | 206.64 g/mol

4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.

Solubility
Soluble in Toluene. Decomposes in water.

Notes
Moisture Sensitive. Store at 4°C. Store in a cool and dark place. Protect from moisture. Store away from strong bases.

Chemical Identifiers

CAS 98-68-0
Molecular Formula C7H7ClO3S
Molecular Weight (g/mol) 206.64
MDL Number MFCD00007446
InChI Key DTJVECUKADWGMO-UHFFFAOYSA-N
Synonym 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy
PubChem CID 7401
IUPAC Name 4-methoxybenzenesulfonyl chloride
SMILES COC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 37°C to 42°C
Boiling Point 119°C to 122°C (0.4 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 609005
Sensitivity Moisture sensitive
Solubility Information Soluble in Toluene. Decomposes in water.
Formula Weight 206.65
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzenesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 202-692-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only