missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzenediazonium Tetrafluoroborate 98.0+%, TCI America™
$285.21 - $989.81
Chemical Identifiers
| CAS | 459-64-3 |
|---|---|
| Molecular Formula | C7H7BF4N2O |
| Molecular Weight (g/mol) | 221.95 |
| MDL Number | MFCD00011897 |
| InChI Key | CACFTKIREZJSIG-UHFFFAOYSA-J |
| Synonym | 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide |
| PubChem CID | 517233 |
| IUPAC Name | boron(3+) 4-methoxybenzene-1-diazonium tetrafluoride |
| SMILES | [B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N |
Chemical Identifiers
| 459-64-3 | |
| 221.95 | |
| CACFTKIREZJSIG-UHFFFAOYSA-J | |
| 517233 | |
| [B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N |
| C7H7BF4N2O | |
| MFCD00011897 | |
| 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide | |
| boron(3+) 4-methoxybenzene-1-diazonium tetrafluoride |
Specifications
| 459-64-3 | |
| C7H7BF4N2O | |
| 5 g | |
| 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide | |
| [B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N | |
| 221.95 | |
| 221.95 | |
| Crystalline Powder |
| 139°C | |
| MFCD00011897 | |
| 1759 | |
| CACFTKIREZJSIG-UHFFFAOYSA-J | |
| boron(3+) 4-methoxybenzene-1-diazonium tetrafluoride | |
| 517233 | |
| ≥98.0% (HPLC) | |
| 4-Methoxybenzenediazonium Tetrafluoroborate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator