missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzamidoxime, 97%

Catalog No. p-7034456
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5214103 1 g
AAH5214106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5214103 Supplier Thermo Scientific Chemicals Supplier No. H5214103
Only null left

CAS: 5373-87-5 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5373-87-5
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD05664434
InChI Key WVALRFKCJCIVBR-UHFFFAOYSA-N
Synonym 4-methoxybenzamidoxime, z-n'-hydroxy-4-methoxybenzimidamide, p-methoxybenzamidoxime, n-hydroxy-4-methoxybenzenecarboximidamide, z-n'-hydroxy-4-methoxybenzene-1-carboximidamide, n-hydroxy-4-methoxy-benzamidine, n'-hydroxy-4-methoxybenzimidamide, hydroxyimino 4-methoxyphenyl methylamine, benzenecarboximidamide, n-hydroxy-4-methoxy, p-methoxybenzamideoxime
PubChem CID 5360053
IUPAC Name N'-hydroxy-4-methoxybenzenecarboximidamide
SMILES COC1=CC=C(C=C1)C(=NO)N

Specifications

Quantity 1 g
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 4-Methoxybenzamidoxime
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
RTECSNumber BZ3425000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only