missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzamide 98.0+%, TCI America™
$121.16 - $121.16
Chemical Identifiers
| CAS | 3424-93-9 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| InChI Key | GUCPYIYFQVTFSI-UHFFFAOYSA-N |
| Synonym | p-anisamide, p-methoxybenzamide, benzamide, 4-methoxy, 4-methoxy benzamide, p-methyoxybenzamide, unii-z85fx3977e, 4-methoxy-benzamide, 4-anisamide, acmc-1cjjo, 4-methoxylbenzamide |
| PubChem CID | 76959 |
| IUPAC Name | 4-methoxybenzamide |
| SMILES | COC1=CC=C(C=C1)C(=O)N |
Chemical Identifiers
| 3424-93-9 | |
| 151.165 | |
| p-anisamide, p-methoxybenzamide, benzamide, 4-methoxy, 4-methoxy benzamide, p-methyoxybenzamide, unii-z85fx3977e, 4-methoxy-benzamide, 4-anisamide, acmc-1cjjo, 4-methoxylbenzamide | |
| 4-methoxybenzamide |
| C8H9NO2 | |
| GUCPYIYFQVTFSI-UHFFFAOYSA-N | |
| 76959 | |
| COC1=CC=C(C=C1)C(=O)N |
Specifications
| 3424-93-9 | |
| 295°C | |
| 5 g | |
| GUCPYIYFQVTFSI-UHFFFAOYSA-N | |
| 4-methoxybenzamide | |
| 76959 | |
| ≥98.0% (GC) | |
| 4-Methoxybenzamide |
| 166°C | |
| C8H9NO2 | |
| p-anisamide, p-methoxybenzamide, benzamide, 4-methoxy, 4-methoxy benzamide, p-methyoxybenzamide, unii-z85fx3977e, 4-methoxy-benzamide, 4-anisamide, acmc-1cjjo, 4-methoxylbenzamide | |
| COC1=CC=C(C=C1)C(=O)N | |
| 151.165 | |
| 151.17 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CV5466666
TSCA : No