missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-o-phenylenediamine, 98%

Catalog No. p-7022672
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1555703 1 g
AAA1555706 5 g
AAA1555714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1555703 Supplier Thermo Scientific Chemicals Supplier No. A1555703
Only null left

CAS: 102-51-2 | C7H10N2O | 138.17 g/mol

4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as a derivatizing reagent by High-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as a derivatizing reagent by High-performance liquid chromatography (HPLC).

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Air sensitive and Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.

Chemical Identifiers

CAS 102-51-2
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00047837
InChI Key AGAHETWGCFCMDK-UHFFFAOYSA-N
Synonym 3,4-diaminoanisole, 4-methoxy-o-phenylenediamine, 4-methoxy-1,2-benzenediamine, 1,2-diamino-4-methoxybenzene, 1,2-benzenediamine, 4-methoxy, 2-amino-4-methoxyphenylamine, 3,4-diamino anisole, o-phenylenediamine, 4-methoxy, 1,2-benzenediamine, 4-methoxy-9ci, 4-methoxy-1,2-diaminobenzene
PubChem CID 153404
IUPAC Name 4-methoxybenzene-1,2-diamine
SMILES COC1=CC(=C(C=C1)N)N

Specifications

Melting Point 46°C to 50°C
Quantity 1 g
UN Number UN2811
Sensitivity Air and light sensitive
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 4-Methoxy-o-phenylenediamine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber ST2690000
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only