missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Methoxy-N-methylformanilide 97.0+%, TCI America™
$290.79 - $898.48
Chemical Identifiers
| CAS | 5279-51-6 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.192 |
| InChI Key | AOXIKOMIXYQQLL-UHFFFAOYSA-N |
| Synonym | N-(4-Methoxyphenyl)-N-methylformamide |
| PubChem CID | 11309718 |
| IUPAC Name | N-(4-methoxyphenyl)-N-methylformamide |
| SMILES | CN(C=O)C1=CC=C(C=C1)OC |
Chemical Identifiers
| 5279-51-6 | |
| 165.192 | |
| N-(4-Methoxyphenyl)-N-methylformamide | |
| N-(4-methoxyphenyl)-N-methylformamide |
| C9H11NO2 | |
| AOXIKOMIXYQQLL-UHFFFAOYSA-N | |
| 11309718 | |
| CN(C=O)C1=CC=C(C=C1)OC |
Specifications
| 5279-51-6 | |
| C9H11NO2 | |
| N-(4-Methoxyphenyl)-N-methylformamide | |
| CN(C=O)C1=CC=C(C=C1)OC | |
| 165.192 | |
| 165.19 | |
| Crystalline Lumps |
| White-Yellow | |
| 1 g | |
| AOXIKOMIXYQQLL-UHFFFAOYSA-N | |
| N-(4-methoxyphenyl)-N-methylformamide | |
| 11309718 | |
| ≥97.0% (GC) | |
| 4′-Methoxy-N-methylformanilide |
Safety and Handling
TSCA : No