missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-N-methylbenzylamine 98.0+%, TCI America™

Catalog No. M28365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28365G 5 g
M28361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28365G Supplier TCI America Supplier No. M28365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 702-24-9
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD04115407
InChI Key AIJFPNKGGAPZFJ-UHFFFAOYSA-N
Synonym 4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine
PubChem CID 485407
IUPAC Name 1-(4-methoxyphenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)OC

Specifications

Color Yellow
Boiling Point 238°C
Quantity 5 g
UN Number 2735
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methoxy-N-methylbenzylamine