missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-alpha-toluenethiol 98.0+%, TCI America™

Catalog No. p-7132570
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11655G 5 g
M116525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11655G Supplier TCI America Supplier No. M11655G
Only null left

Chemical Identifiers

CAS 6258-60-2
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00004871
InChI Key PTDVPWWJRCOIIO-UHFFFAOYSA-N
Synonym 4-methoxybenzyl mercaptan, 4-methoxy-alpha-toluenethiol, 4-methoxybenzylmercaptan, 4-methoxyphenyl methanethiol, 4-methoxy benzylmercaptan, 4-methoxybenzenemethanethiol, benzenemethanethiol, 4-methoxy, p-methoxybenzyl mercaptan, p-methoxybenzylthiol, p-methoxybenzylmercaptan
PubChem CID 80407
IUPAC Name (4-methoxyphenyl)methanethiol
SMILES COC1=CC=C(CS)C=C1

Specifications

Color Yellow
Boiling Point 126°C
Quantity 5 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxy-alpha-toluenethiol