missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-alpha-toluenethiol, 95%, Thermo Scientific Chemicals

Catalog No. p-3779006
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150950250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC150950250 Supplier Thermo Scientific Chemicals Supplier No. 150950250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6258-60-2 | Molecular Formula: C8H10OS | Molecular Weight: 154.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6258-60-2
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00004871
InChI Key PTDVPWWJRCOIIO-UHFFFAOYSA-N
Synonym 4-methoxybenzyl mercaptan, 4-methoxy-alpha-toluenethiol, 4-methoxybenzylmercaptan, 4-methoxyphenyl methanethiol, 4-methoxy benzylmercaptan, 4-methoxybenzenemethanethiol, benzenemethanethiol, 4-methoxy, p-methoxybenzyl mercaptan, p-methoxybenzylthiol, p-methoxybenzylmercaptan
PubChem CID 80407
IUPAC Name (4-methoxyphenyl)methanethiol
SMILES COC1=CC=C(CS)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1070g/mL
Boiling Point 90°C to 95°C (0.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5720 to 1.578
Linear Formula CH3OC6H4CH2SH
Quantity 25 g
Beilstein 06, 901
Specific Gravity 1.107
Solubility Information Solubility in water: insoluble
Formula Weight 154.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Methoxy-α-toluenethiol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 228-393-6

RUO – Research Use Only