missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-6-methyl-2H-pyran-2-one, 97%, Thermo Scientific Chemicals

Catalog No. aah6139503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6139503 1 g
AAH61395MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6139503 Supplier Thermo Scientific Chemicals Supplier No. H6139503

4-Methoxy-6-methyl-2H-pyran-2-one is used as a model compound to study the assignment of ring proton resonances in 4-methoxy-6-nonyl-2H-pyran-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-6-methyl-2H-pyran-2-one is used as a model compound to study the assignment of ring proton resonances in 4-methoxy-6-nonyl-2H-pyran-2-one.

Solubility
Poorly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 672-89-9
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00006640
InChI Key MTZAUZNQAMNFME-UHFFFAOYSA-N
Synonym 4-methoxy-6-methyl-2h-pyran-2-one, methyltriacetolactone, methyltriacetolacton, 2h-pyran-2-one, 4-methoxy-6-methyl, 4-methoxy-6-methyl-pyran-2-one, methyltriacetolacton german, 4-methoxy-6-methyl-2-pyrone, 4mhp, wln: t6ovj do1 f1, 4-methoxy-6-methyl-2-pyranone
PubChem CID 12651
IUPAC Name 4-methoxy-6-methylpyran-2-one
SMILES COC1=CC(=O)OC(C)=C1

Specifications

Melting Point 83°C to 86°C
Quantity 1 g
Solubility Information Poorly soluble in water.
Formula Weight 140.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Methoxy-6-methyl-2H-pyran-2-one
EINECSNumber 211-600-9
RTECSNumber UQ1225500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only