CAS RN 672-89-9
IUPAC Name:
4-methoxy-6-methyl-2H-pyran-2-one
Synonyms:
4-methoxy-6-methylpyran-2-one
Molecular Weight (g/mol):
140.14
Molecular Formula:
C7H8O3
InChi Key:
MTZAUZNQAMNFME-UHFFFAOYSA-N
SMILES:
COC1=CC(=O)OC(C)=C1
1
–
1
of
1
results
1
Promotions Available
4-Methoxy-6-methyl-2H-pyran-2-one, 97%, Thermo Scientific Chemicals
4-Methoxy-6-methyl-2H-pyran-2-one, 97%, Thermo Scientific Chemicals
CAS: 672-89-9 Molecular Formula: C7H8O3 Molecular Weight (g/mol): 140.14 MDL Number: MFCD00006640 InChI Key: MTZAUZNQAMNFME-UHFFFAOYSA-N Synonym: 4-methoxy-6-methyl-2h-pyran-2-one,methyltriacetolactone,methyltriacetolacton,2h-pyran-2-one, 4-methoxy-6-methyl,4-methoxy-6-methyl-pyran-2-one,methyltriacetolacton german,4-methoxy-6-methyl-2-pyrone,4mhp,wln: t6ovj do1 f1,4-methoxy-6-methyl-2-pyranone PubChem CID: 12651 IUPAC Name: 4-methoxy-6-methylpyran-2-one SMILES: COC1=CC(=O)OC(C)=C1