missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-4'-nitrobiphenyl 98.0+%, TCI America™
$53.90 - $317.66
Chemical Identifiers
| CAS | 2143-90-0 |
|---|---|
| Molecular Formula | C13H11NO3 |
| Molecular Weight (g/mol) | 229.235 |
| MDL Number | MFCD00143334 |
| InChI Key | AUIINENVMPWGQF-UHFFFAOYSA-N |
| Synonym | 4-(4′C-Methoxyphenyl)nitrobenzene, 4-(4′C-Nitrophenyl)anisole |
| PubChem CID | 11128079 |
| IUPAC Name | 1-(4-methoxyphenyl)-4-nitrobenzene |
| SMILES | COC1=CC=C(C=C1)C2=CC=C(C=C2)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M0997100MG
|
TCI America
M0997100MG |
100 mg |
Each for $53.90
|
|
|||||
|
M09971G
|
TCI America
M09971G |
1 g |
Each for $317.66
|
|
|||||
Chemical Identifiers
| 2143-90-0 | |
| 229.235 | |
| AUIINENVMPWGQF-UHFFFAOYSA-N | |
| 11128079 | |
| COC1=CC=C(C=C1)C2=CC=C(C=C2)[N+](=O)[O-] |
| C13H11NO3 | |
| MFCD00143334 | |
| 4-(4′C-Methoxyphenyl)nitrobenzene, 4-(4′C-Nitrophenyl)anisole | |
| 1-(4-methoxyphenyl)-4-nitrobenzene |
Specifications
| 2143-90-0 | |
| Yellow | |
| MFCD00143334 | |
| 4-(4′C-Methoxyphenyl)nitrobenzene, 4-(4′C-Nitrophenyl)anisole | |
| COC1=CC=C(C=C1)C2=CC=C(C=C2)[N+](=O)[O-] | |
| 229.235 | |
| 229.24 | |
| Crystalline Powder |
| 110°C | |
| C13H11NO3 | |
| 100 mg | |
| AUIINENVMPWGQF-UHFFFAOYSA-N | |
| 1-(4-methoxyphenyl)-4-nitrobenzene | |
| 11128079 | |
| ≥98.0% (GC) | |
| 4-Methoxy-4′-nitrobiphenyl |
Safety and Handling
TSCA : No