accessibility menu, dialog, popup

UserName

H3CONOO

CAS RN 2143-90-0

IUPAC Name: 4-methoxy-4'-nitro-1,1'-biphenyl
Synonyms: 4-methoxy-4'-nitro-1,1'-biphenyl
Molecular Weight (g/mol): 229.24
Molecular Formula: C13H11NO3
InChi Key: AUIINENVMPWGQF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)