missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-4'-nitrobiphenyl 98.0+%, TCI America™

Catalog No. M09971G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09971G 1 g
M0997100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09971G Supplier TCI America Supplier No. M09971G
Only null left

Chemical Identifiers

CAS 2143-90-0
Molecular Formula C13H11NO3
Molecular Weight (g/mol) 229.235
MDL Number MFCD00143334
InChI Key AUIINENVMPWGQF-UHFFFAOYSA-N
Synonym 4-(4′C-Methoxyphenyl)nitrobenzene, 4-(4′C-Nitrophenyl)anisole
PubChem CID 11128079
IUPAC Name 1-(4-methoxyphenyl)-4-nitrobenzene
SMILES COC1=CC=C(C=C1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 110°C
Color Yellow
Quantity 1 g
Formula Weight 229.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-4′-nitrobiphenyl