missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-4-methyl-2-pentanone 96.0+%, TCI America™

Catalog No. M065225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M065225ML 25 mL
M0652500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M065225ML Supplier TCI America Supplier No. M065225ML

Chemical Identifiers

CAS 107-70-0
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00043932
InChI Key KOKPBCHLPVDQTK-UHFFFAOYSA-N
Synonym 4-methoxy-4-methyl-2-pentanone, pentoxone, pent-oxone solvent, 2-pentanone, 4-methoxy-4-methyl, 4-methyl-4-methoxy-2-pentanone, me 6 k, ccris 4840, acmc-1bwvk, dsstox_cid_5557, dsstox_rid_77828
PubChem CID 7884
IUPAC Name 4-methoxy-4-methylpentan-2-one
SMILES CC(=O)CC(C)(C)OC

Specifications

Color Colorless
Boiling Point 156°C
Quantity 25 mL
UN Number 2293
Formula Weight 130.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxy-4-methyl-2-pentanone
EINECSNumber (2)-0585
RTECSNumber SA9185000
TSCA Yes