missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. M30955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30955G 5 g
M309525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30955G Supplier TCI America Supplier No. M30955G

Chemical Identifiers

CAS 213598-09-5
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.15
MDL Number MFCD01091014
InChI Key VNWUPESTVBTYJM-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzoic acid, 4-methoxy-3-trifluoromethyl-benzoic acid, acmc-1cr7h, 3-trifluoromethyl-4-methoxybenzoic acid, benzoic acid, 4-methoxy-3-trifluoromethyl, 4-methyloxy-3-trifluoromethyl benzoic acid
PubChem CID 2775295
IUPAC Name 4-methoxy-3-(trifluoromethyl)benzoic acid
SMILES COC1=CC=C(C=C1C(F)(F)F)C(O)=O

Specifications

Melting Point 172°C
Quantity 5 g
Formula Weight 220.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzoic Acid