missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-3-nitrobiphenyl 99.0+%, TCI America™
$53.90 - $180.97
Chemical Identifiers
| CAS | 15854-73-6 |
|---|---|
| Molecular Formula | C13H11NO3 |
| Molecular Weight (g/mol) | 229.24 |
| MDL Number | MFCD00191554 |
| InChI Key | HVYOIIIEAVPMCR-UHFFFAOYSA-N |
| Synonym | 2-Nitro-4-phenylanisole |
| PubChem CID | 615238 |
| IUPAC Name | 4-methoxy-3-nitro-1,1'-biphenyl |
| SMILES | COC1=C(C=C(C=C1)C1=CC=CC=C1)[N+]([O-])=O |
Chemical Identifiers
| 15854-73-6 | |
| 229.24 | |
| HVYOIIIEAVPMCR-UHFFFAOYSA-N | |
| 615238 | |
| COC1=C(C=C(C=C1)C1=CC=CC=C1)[N+]([O-])=O |
| C13H11NO3 | |
| MFCD00191554 | |
| 2-Nitro-4-phenylanisole | |
| 4-methoxy-3-nitro-1,1'-biphenyl |
Specifications
| 15854-73-6 | |
| Yellow | |
| MFCD00191554 | |
| 2-Nitro-4-phenylanisole | |
| COC1=C(C=C(C=C1)C1=CC=CC=C1)[N+]([O-])=O | |
| 229.24 | |
| 229.24 | |
| Crystalline Powder |
| 89°C | |
| C13H11NO3 | |
| 5 g | |
| HVYOIIIEAVPMCR-UHFFFAOYSA-N | |
| 4-methoxy-3-nitro-1,1'-biphenyl | |
| 615238 | |
| ≥99.0% (GC) | |
| 4-Methoxy-3-nitrobiphenyl |
Safety and Handling
TSCA : No