missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-nitrobiphenyl 99.0+%, TCI America™

Catalog No. M11665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11665G 5 g
M116625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11665G Supplier TCI America Supplier No. M11665G
Only null left

Chemical Identifiers

CAS 15854-73-6
Molecular Formula C13H11NO3
Molecular Weight (g/mol) 229.24
MDL Number MFCD00191554
InChI Key HVYOIIIEAVPMCR-UHFFFAOYSA-N
Synonym 2-Nitro-4-phenylanisole
PubChem CID 615238
IUPAC Name 4-methoxy-3-nitro-1,1'-biphenyl
SMILES COC1=C(C=C(C=C1)C1=CC=CC=C1)[N+]([O-])=O

Specifications

Melting Point 89°C
Color Yellow
Quantity 5 g
Formula Weight 229.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3-nitrobiphenyl