missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-nitrobenzaldehyde, 98%

Catalog No. p-7023486
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1708106 5 g
AAA1708114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1708106 Supplier Thermo Scientific Chemicals Supplier No. A1708106

CAS: 31680-08-7 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31680-08-7
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00126498
InChI Key YTCRQCGRYCKYNO-UHFFFAOYSA-N
Synonym 3-nitro-4-anisaldehyde, 4-methoxy-3-nitro-benzaldehyde, benzaldehyde, 4-methoxy-3-nitro, 3-nitro-4-methoxybenzaldehyde, pubchem8145, 3-nitro-p-anisaldehyde, acmc-1adwt, 4-methoxy-5-nitrobenzaldehyde, 4-methoxy-3-nitrobenzaldehyde
PubChem CID 700608
IUPAC Name 4-methoxy-3-nitrobenzaldehyde
SMILES COC1=CC=C(C=O)C=C1[N+]([O-])=O

Specifications

Melting Point 97°C to 100°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 181.15
Percent Purity 98%
Chemical Name or Material 4-Methoxy-3-nitrobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only