missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3,5-dimethylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7133046
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21391G 1 g
M21395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21391G Supplier TCI America Supplier No. M21391G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21553-46-8
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00020309
InChI Key WXVQURJGDUNJCS-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-methoxybenzoic acid, 3,5-dimethyl-p-anisic acid, 4-methoxy-3,5-dimethyl-benzoic acid, 3,5-dimethyl anisic acid, benzoic acid, 4-methoxy-3,5-dimethyl, 4-methoxy-3,5-dimethylbenzoicacid, acmc-209flo, ksc496g8f, 3,5-dimethyl-4-methoxy-benzoic acid, 4-methoxy-3,5-dimethyl benzoic acid
PubChem CID 88944
IUPAC Name 4-methoxy-3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1OC)C)C(=O)O

Specifications

Melting Point 194°C
Color Yellow
Quantity 1 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3,5-dimethylbenzoic Acid