missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2-nitrobenzenesulfonyl Chloride 93.0+%, TCI America™
Specifications
Specifications
| Melting Point | 77°C |
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 251.64 |
| Percent Purity | ≥93.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methoxy-2-nitrobenzenesulfonyl Chloride |
Safety and Handling
| TSCA | No |