missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-nitroaniline 98.0+%, TCI America™

Catalog No. p-7132002
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M011925G 25 g
M0119100G 100 g
M0119500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M011925G Supplier TCI America Supplier No. M011925G
Only null left

Chemical Identifiers

CAS 96-96-8
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD00007152
InChI Key QFMJFXFXQAFGBO-UHFFFAOYSA-N
Synonym 2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt
PubChem CID 66793
ChEBI CHEBI:48973
IUPAC Name 4-methoxy-2-nitroaniline
SMILES COC1=CC(=C(C=C1)N)[N+](=O)[O-]

Specifications

Melting Point 124°C
Quantity 25 g
UN Number 2811
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2-nitroaniline
EINECSNumber (5)-2296
RTECSNumber BY4415000
TSCA Yes