Learn More
4-Methoxy-2-nitroaniline, 99%
CAS: 96-96-8 | C7H8N2O3 | 168.152 g/mol
Supplier: Thermo Scientific Chemicals A116730E
Description
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.
Solubility
Slightly soluble in water. Soluble in hot methanol almost transparency.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications
| 4-Methoxy-2-nitroaniline | |
| 126°C to 128°C | |
| MFCD00007152 | |
| UN2811 | |
| 2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt | |
| QFMJFXFXQAFGBO-UHFFFAOYSA-N | |
| 4-methoxy-2-nitroaniline | |
| 66793 | |
| 168.15 |
| 96-96-8 | |
| C7H8N2O3 | |
| 2500 g | |
| 880318 | |
| Slightly soluble in water. Soluble in hot methanol almost transparency. | |
| COC1=CC(=C(C=C1)N)[N+](=O)[O-] | |
| 168.152 | |
| CHEBI:48973 | |
| 99% |
Chemical Identifiers
| 96-96-8 | |
| 168.152 | |
| QFMJFXFXQAFGBO-UHFFFAOYSA-N | |
| 66793 | |
| 4-methoxy-2-nitroaniline |
| C7H8N2O3 | |
| MFCD00007152 | |
| 2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt | |
| CHEBI:48973 | |
| COC1=CC(=C(C=C1)N)[N+](=O)[O-] |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 202-547-2
RTECSNumber : BY4415000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only