missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7133421
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27801G 1 g
M27805G 5 g
M278025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M27801G Supplier TCI America Supplier No. M27801G
Only null left

Chemical Identifiers

CAS 6245-57-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00020291
InChI Key MSVRGYOYISBGTH-UHFFFAOYSA-N
Synonym 2-methyl-4-methoxybenzoic acid, 4-methoxy-2-methyl-benzoic acid, 2-methyl-p-anisic acid, benzoic acid, 4-methoxy-2-methyl, 4-methoxy-o-toluic acid, acmc-209x9h, ksc495k2d, 2-methyl-4-methoxybenzoicacid, 2-methyl-4-methoxy-benzoic acid, 4-methoxy-2-methyl benzoic acid
PubChem CID 597216
IUPAC Name 4-methoxy-2-methylbenzoic acid
SMILES COC1=CC(C)=C(C=C1)C(O)=O

Specifications

Melting Point 180°C
Color White
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2-methylbenzoic Acid