missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylbenzaldehyde, 95%

Catalog No. p-7055658
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6429503 1 g
AAH6429506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6429503 Supplier Thermo Scientific Chemicals Supplier No. H6429503

CAS: 52289-54-0 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52289-54-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD02261771
InChI Key WICYVKGMEJSDAO-UHFFFAOYSA-N
Synonym 2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
PubChem CID 283285
IUPAC Name 4-methoxy-2-methylbenzaldehyde
SMILES CC1=C(C=CC(=C1)OC)C=O

Specifications

Color Yellow
Density 1.107 g/mL
Boiling Point 262°C to 267°C (literature)
Flash Point >110°C (230°F)
Refractive Index 1.571
Quantity 1 g
Beilstein 1933786
Formula Weight 150.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Methoxy-2-methylbenzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only