missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylaniline 99.0+%, TCI America™

Catalog No. p-7132754
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M147425G 25 g
M1474500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M147425G Supplier TCI America Supplier No. M147425G

Chemical Identifiers

CAS 102-50-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007735
InChI Key CDGNLUSBENXDGG-UHFFFAOYSA-N
Synonym m-cresidine, 2-methyl-4-methoxyaniline, 2-methyl-p-anisidine, 4-methoxy-o-toluidine, benzenamine, 4-methoxy-2-methyl, meta-cresidine, p-anisidine, 2-methyl, 6-amino-3-methoxytoluene, 2-methyl-4-methoxybenzenamine, 4-methoxy-2-methylbenzenamine
PubChem CID 7610
ChEBI CHEBI:82430
IUPAC Name 4-methoxy-2-methylaniline
SMILES CC1=C(C=CC(=C1)OC)N

Specifications

Melting Point 13°C
Boiling Point 249°C
Quantity 25 g
UN Number 2810
Formula Weight 137.18
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methoxy-2-methylaniline
EINECSNumber (3)-0614
RTECSNumber BZ6730000
TSCA Yes
Recommended Storage Refrigerator