missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2,3,6-trimethylbenzaldehyde 98.0+%, TCI America™

Catalog No. M30025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30025G 5 g
M30021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30025G Supplier TCI America Supplier No. M30025G
Only null left

Chemical Identifiers

CAS 54344-92-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00456729
InChI Key BTOFIDLWQJCUJG-UHFFFAOYSA-N
Synonym 2,3,6-Trimethyl-p-anisaldehyde
PubChem CID 824142
IUPAC Name 4-methoxy-2,3,6-trimethylbenzaldehyde
SMILES CC1=CC(=C(C(=C1C=O)C)C)OC

Specifications

Melting Point 66°C
Color Yellow
Quantity 5 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2,3,6-trimethylbenzaldehyde
TSCA No
Recommended Storage Freezer (-20 °C)