missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-1-naphthoic Acid 98.0+%, TCI America™

Catalog No. M27635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27635G 5 g
M27631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27635G Supplier TCI America Supplier No. M27635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13041-62-8
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD02575372
InChI Key WRQHSQDGYYDRMX-UHFFFAOYSA-N
Synonym 4-methoxy-1-naphthoic acid, 4-methoxy-naphthalene-1-carboxylic acid, 1-naphthalenecarboxylic acid, 4-methoxy, 1-naphthalenecarboxylicacid, 4-methoxy, 4-methoxynaphthalenecarboxylic acid, ksc181m4d, 4-methoxy-1-naphthalenecarboxylic acid
PubChem CID 765380
IUPAC Name 4-methoxynaphthalene-1-carboxylic acid
SMILES COC1=CC=C(C2=CC=CC=C21)C(=O)O

Specifications

Melting Point 245°C
Quantity 5 g
Formula Weight 202.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-1-naphthoic Acid