missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 89°C |
| Quantity | 1 g |
| Formula Weight | 240.32 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |