missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Mercaptopyridine 97.0+%, TCI America™
Supplier: TCI America M063025G
Specifications
| 4-Mercaptopyridine | |
| 180°C | |
| MFCD00006422 | |
| 4-mercaptopyridine, pyridine-4-thiol, 4-pyridinethiol, 4-thiopyridine, 4-pyridinethione, pyridine, 4-mercapto, 4-thiopyridone, 4 1h-pyridinethione, pyridine-4 1h-thione, 5-21-07-00167 beilstein handbook reference | |
| S=C1C=CNC=C1 | |
| 111.16 | |
| 111.16 | |
| Crystalline Powder |
| 4556-23-4 | |
| C5H5NS | |
| 25 g | |
| FHTDDANQIMVWKZ-UHFFFAOYSA-N | |
| 1,4-dihydropyridine-4-thione | |
| 2723889 | |
| ≥97.0% (T) |
Chemical Identifiers
| 4556-23-4 | |
| 111.16 | |
| FHTDDANQIMVWKZ-UHFFFAOYSA-N | |
| 2723889 | |
| S=C1C=CNC=C1 |
| C5H5NS | |
| MFCD00006422 | |
| 4-mercaptopyridine, pyridine-4-thiol, 4-pyridinethiol, 4-thiopyridine, 4-pyridinethione, pyridine, 4-mercapto, 4-thiopyridone, 4 1h-pyridinethione, pyridine-4 1h-thione, 5-21-07-00167 beilstein handbook reference | |
| 1,4-dihydropyridine-4-thione |
Safety and Handling
RTECSNumber : UT8578000
TSCA : No
Recommended Storage : Freezer