missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptophenylacetic acid, 97%

Catalog No. AAH2765803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2765803 1 g
AAH2765806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2765803 Supplier Thermo Scientific Chemicals Supplier No. H2765803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39161-84-7 | C8H8O2S | 168.21 g/mol

An useful synthetic intermediate for the preparation of novel anti inflammatory agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.

Solubility
Slightly soluble in water. Soluble in Dichloromethane, Ethanol and Methanol

Notes
Air sensitive. Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 39161-84-7
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00797617
InChI Key ORXSLDYRYTVAPC-UHFFFAOYSA-N
Synonym 4-mercaptophenylacetic acid, 2-4-mercaptophenyl acetic acid, 2-4-sulfanylphenyl acetic acid, benzeneacetic acid, 4-mercapto, 4-mercapto-phenyl-acetic acid, acmc-20amfq, 4-mercaptobenzeneacetate, 4-mercapto-phenylacetic acid, 4-mercaptophenyl acetic acid, 4-mercapto-phenyl acetic acid
PubChem CID 3390511
IUPAC Name 2-(4-sulfanylphenyl)acetic acid
SMILES OC(=O)CC1=CC=C(S)C=C1

Specifications

Melting Point 105°C to 110°C
Quantity 1 g
Sensitivity Air sensitive
Solubility Information Slightly soluble in water. Soluble in Dichloromethane,Ethanol and Methanol
Formula Weight 168.21
Percent Purity 97%
Chemical Name or Material 4-Mercaptophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only