missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptohydrocinnamic Acid 95.0+%, TCI America™

Catalog No. M09175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09175G 5 g
M09171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09175G Supplier TCI America Supplier No. M09175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63545-55-1
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.24
MDL Number MFCD00060121
InChI Key YQWPHBFLHAJVCG-UHFFFAOYSA-N
Synonym 3-(4-Mercaptophenyl)propionic Acid
PubChem CID 579784
IUPAC Name 3-(4-sulfanylphenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(S)C=C1

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 5 g
Formula Weight 182.24
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Mercaptohydrocinnamic Acid
TSCA No
Recommended Storage Refrigerator