accessibility menu, dialog, popup

UserName

OHOSH

CAS RN 63545-55-1

IUPAC Name: 3-(4-sulfanylphenyl)propanoic acid
Synonyms: 3-(4-sulfanylphenyl)propanoic acid
Molecular Weight (g/mol): 182.24
Molecular Formula: C9H10O2S
InChi Key: YQWPHBFLHAJVCG-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(S)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results