missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptobenzoic acid, 90%

Catalog No. p-4089562
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC373300050 5 g Glass bottle
AC373300250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC373300050 Supplier Thermo Scientific Chemicals Supplier No. 373300050
Only null left

CAS: 1074-36-8 | C7H6O2S | 154.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1074-36-8
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00016617
InChI Key LMJXSOYPAOSIPZ-UHFFFAOYSA-N
Synonym 4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol
PubChem CID 95738
IUPAC Name 4-sulfanylbenzoic acid
SMILES OC(=O)C1=CC=C(S)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 223.0°C to 225.0°C
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Linear Formula HSC6H4CO2H
Quantity 5 g
Formula Weight 154.19
Percent Purity 90%
Chemical Name or Material 4-Mercaptobenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if

RUO – Research Use Only