Learn More
Agilent BioTek 4 L Waste Bottle with Level Sense
Waste Bottle for use with various microplate washers.
Supplier:  Agilent BioTek 7100542
Catalog No. BT7100542
Only null left
Explore available promotions
Description
Description
  • For use with 405 TS, 405 LS, ELx405 Select Deep Well, EL406 and 406 FX
Specifications
Specifications
4 L
Waste Bottle
405 TS Microplate Washer, 405 LS Microplate Washer, EL406 Microplate Washer, 406 FX Microplate Washer
Product Suggestions
Product Suggestions