missing translation for 'onlineSavingsMsg'
Learn More

4-Ketopimelic acid, 98%

Catalog No. p-3778914
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122880250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC122880250 Supplier Thermo Scientific Chemicals Supplier No. 122880250

CAS: 502-50-1 | C7H10O5 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 502-50-1
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00002798
InChI Key UDDSEESQRGPVIL-UHFFFAOYSA-N
PubChem CID 95084
IUPAC Name 4-oxoheptanedioic acid
SMILES OC(=O)CCC(=O)CCC(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 142.0°C to 144.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2CH2COCH2CH2CO2H
Quantity 25 g
Beilstein 03, 804
Synonym 4-ketopimelic acid, 4-oxopimelic acid, 4-ketopimelic, .gamma.-ketopimelic acid, heptanedioic acid, 4-oxo, acmc-20amfs, pimelic acid, 4-keto, heptanedioic acid,4-oxo, 4-oxoheptanedioic acid
Formula Weight 174.15
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 4-Ketopimelic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-941-8

RUO – Research Use Only