accessibility menu, dialog, popup

UserName

CAS RN 502-50-1

IUPAC Name: 4-oxoheptanedioic acid
Synonyms: 4-oxoheptanedioic acid
Molecular Weight (g/mol): 174.15
Molecular Formula: C7H10O5
InChi Key: UDDSEESQRGPVIL-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)CCC(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)