missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. I06205G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06205G Supplier TCI America Supplier No. I06205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Isopropylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 16152-51-5
Melting Point 140°C
Color White-Yellow
Molecular Formula C9H13BO2
Quantity 5 g
MDL Number MFCD01074614
Synonym 4-isopropylphenylboronic acid, 4-isopropylbenzeneboronic acid, 4-isopropylphenyl boronic acid, 4-propan-2-ylphenyl boronic acid, p-isopropylphenylboronic acid, p-cumylboronic acid, 4-cumylboronic acid, 4-iso-propylphenyl boronic acid, 4-iso-propylphenylboronic acid
InChI Key IAEUFBDMVKQCLU-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C(C)C)(O)O
IUPAC Name (4-propan-2-ylphenyl)boronic acid
Molecular Weight (g/mol) 164.011
PubChem CID 2773480
Formula Weight 164.01
Physical Form Crystalline Powder
Show More Show Less