missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenylacetic acid, 98+%

Catalog No. p-7044185
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0507103 1 g
AAL0507106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0507103 Supplier Thermo Scientific Chemicals Supplier No. L0507103

CAS: 4476-28-2 | C11H14O2 | 178.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4476-28-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00041035
InChI Key RERBQXVRXYCGLT-UHFFFAOYSA-N
Synonym 4-isopropylphenylacetic acid, 2-4-isopropylphenyl acetic acid, 4-isopropylphenyl acetic acid, benzeneacetic acid, 4-1-methylethyl, 2-4-propan-2-yl phenyl acetic acid, p-isopropyl phenyl acetic acid, 4-isopropyl-phenyl-acetic acid, 4-propan-2-yl phenyl acetic acid, 2-4-methylethyl phenyl acetic acid, 4-isopropylphenylaceticacid
PubChem CID 78230
IUPAC Name 2-(4-propan-2-ylphenyl)acetic acid
SMILES CC(C)C1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 51°C to 52°C
Quantity 1 g
Beilstein 2328756
Formula Weight 178.23
Percent Purity ≥98%
Chemical Name or Material 4-Isopropylphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-755-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only