accessibility menu, dialog, popup

UserName

CAS RN 4476-28-2

IUPAC Name: 2-[4-(propan-2-yl)phenyl]acetate
Synonyms: 2-(4-isopropylphenyl)acetate
Molecular Weight (g/mol): 177.22
Molecular Formula: C11H13O2
InChi Key: RERBQXVRXYCGLT-UHFFFAOYSA-M
SMILES: CC(C)C1=CC=C(CC([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results